Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601937 MIM: 610013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCOA3 - nuclear receptor coactivator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULF2 - sulfatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161841.1 | Intron | NP_001155313.1 | ||||
NM_018837.3 | Intron | NP_061325.1 | ||||
NM_198596.2 | Intron | NP_940998.2 | ||||
XM_005260458.2 | Intron | XP_005260515.1 | ||||
XM_006723827.1 | Intron | XP_006723890.1 | ||||
XM_006723828.1 | Intron | XP_006723891.1 | ||||
XM_006723829.2 | Intron | XP_006723892.1 | ||||
XM_006723830.1 | Intron | XP_006723893.1 | ||||
XM_011528913.1 | Intron | XP_011527215.1 | ||||
XM_011528914.1 | Intron | XP_011527216.1 | ||||
XM_011528915.1 | Intron | XP_011527217.1 | ||||
XM_011528916.1 | Intron | XP_011527218.1 | ||||
XM_017027955.1 | Intron | XP_016883444.1 |