Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATTTCTGTTGTTCCCAGAGCGA[A/G]GGGATGAGAAGCCTGAGGCCGGCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602686 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100127955 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100127955 - uncharacterized LOC100127955 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAD1L1 - MAD1 mitotic arrest deficient like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013836.1 | Intron | NP_001013858.1 | ||||
NM_001013837.1 | Intron | NP_001013859.1 | ||||
NM_001304523.1 | Intron | NP_001291452.1 | ||||
NM_001304524.1 | Intron | NP_001291453.1 | ||||
NM_001304525.1 | Intron | NP_001291454.1 | ||||
NM_003550.2 | Intron | NP_003541.2 | ||||
XM_005249877.1 | Intron | XP_005249934.1 | ||||
XM_011515567.1 | Intron | XP_011513869.1 | ||||
XM_011515568.2 | Intron | XP_011513870.1 | ||||
XM_011515569.2 | Intron | XP_011513871.2 | ||||
XM_011515571.1 | Intron | XP_011513873.1 | ||||
XM_017012690.1 | Intron | XP_016868179.1 | ||||
XM_017012691.1 | Intron | XP_016868180.1 |
MIR4655 - microRNA 4655 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |