Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616773 MIM: 613121 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIGA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIGA1 - mitoguardin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEXN - nexilin F-actin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172309.1 | Intron | NP_001165780.1 | ||||
NM_144573.3 | Intron | NP_653174.3 | ||||
XM_005271322.3 | Intron | XP_005271379.1 | ||||
XM_005271323.3 | Intron | XP_005271380.1 | ||||
XM_005271324.4 | Intron | XP_005271381.1 | ||||
XM_005271325.3 | Intron | XP_005271382.1 | ||||
XM_005271326.3 | Intron | XP_005271383.1 | ||||
XM_005271327.3 | Intron | XP_005271384.1 |
NEXN-AS1 - NEXN antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |