Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C20orf197 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C20orf197 - chromosome 20 open reading frame 197 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302813.1 | Intron | NP_001289742.1 | ||||
NM_001302815.1 | Intron | NP_001289744.1 | ||||
NM_173644.2 | Intron | NP_775915.1 | ||||
XM_017027828.1 | Intron | XP_016883317.1 | ||||
XM_017027829.1 | Intron | XP_016883318.1 | ||||
XM_017027830.1 | Intron | XP_016883319.1 | ||||
XM_017027831.1 | Intron | XP_016883320.1 | ||||
XM_017027832.1 | Intron | XP_016883321.1 |