Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616826 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS15L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EPS15L1 - epidermal growth factor receptor pathway substrate 15 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258374.1 | Intron | NP_001245303.1 | ||||
NM_001258375.1 | Intron | NP_001245304.1 | ||||
NM_001258376.1 | Intron | NP_001245305.1 | ||||
NM_021235.2 | Intron | NP_067058.1 | ||||
XM_017027086.1 | Intron | XP_016882575.1 | ||||
XM_017027087.1 | Intron | XP_016882576.1 | ||||
XM_017027088.1 | Intron | XP_016882577.1 | ||||
XM_017027089.1 | Intron | XP_016882578.1 | ||||
XM_017027090.1 | Intron | XP_016882579.1 | ||||
XM_017027091.1 | Intron | XP_016882580.1 | ||||
XM_017027092.1 | Intron | XP_016882581.1 |
Set Membership: |
HapMap |