Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606078 MIM: 610440 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MKL1 - megakaryoblastic leukemia (translocation) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282660.1 | Intron | NP_001269589.1 | ||||
NM_001282661.1 | Intron | NP_001269590.1 | ||||
NM_001282662.1 | Intron | NP_001269591.1 | ||||
NM_001318139.1 | Intron | NP_001305068.1 | ||||
NM_020831.4 | Intron | NP_065882.1 | ||||
XM_005261692.1 | Intron | XP_005261749.1 | ||||
XM_005261694.1 | Intron | XP_005261751.1 | ||||
XM_011530283.1 | Intron | XP_011528585.1 | ||||
XM_011530285.1 | Intron | XP_011528587.1 | ||||
XM_011530287.1 | Intron | XP_011528589.1 | ||||
XM_017028888.1 | Intron | XP_016884377.1 | ||||
XM_017028889.1 | Intron | XP_016884378.1 |
SGSM3 - small G protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |