Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600816 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSPA8 - heat shock protein family A (Hsp70) member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006597.5 | Intron | NP_006588.1 | ||||
NM_153201.3 | Intron | NP_694881.1 | ||||
XM_011542798.1 | Intron | XP_011541100.1 |
SNORD14C - small nucleolar RNA, C/D box 14C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD14D - small nucleolar RNA, C/D box 14D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD14E - small nucleolar RNA, C/D box 14E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |