Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606735 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OSBPL7 - oxysterol binding protein like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145798.2 | 2980 | Silent Mutation | TAC,TAT | Y,Y 833 | NP_665741.1 | |
XM_017024122.1 | 2980 | Silent Mutation | TAC,TAT | Y,Y 879 | XP_016879611.1 | |
XM_017024123.1 | 2980 | Silent Mutation | TAC,TAT | Y,Y 879 | XP_016879612.1 | |
XM_017024124.1 | 2980 | Silent Mutation | TAC,TAT | Y,Y 861 | XP_016879613.1 | |
XM_017024125.1 | 2980 | Silent Mutation | TAC,TAT | Y,Y 441 | XP_016879614.1 |