Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACTTTATTTGCTCATGATGCAG[G/T]TTAAGCCCAGTTTCTGGCATGTGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf64 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C5orf64 - chromosome 5 open reading frame 64 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173667.3 | Intron | NP_775938.1 | ||||
XM_017009378.1 | Intron | XP_016864867.1 | ||||
XM_017009379.1 | Intron | XP_016864868.1 | ||||
XM_017009380.1 | Intron | XP_016864869.1 | ||||
XM_017009381.1 | Intron | XP_016864870.1 | ||||
XM_017009382.1 | Intron | XP_016864871.1 |
Set Membership: |
HapMap |