Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 602626 | |||||||||||||||||||||||
Literature Links: |
IL1RAP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
IL1RAP - interleukin 1 receptor accessory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167928.1 | Intron | NP_001161400.1 | ||||
NM_001167929.1 | Intron | NP_001161401.1 | ||||
NM_001167930.1 | Intron | NP_001161402.1 | ||||
NM_001167931.1 | Intron | NP_001161403.1 | ||||
NM_002182.3 | Intron | NP_002173.1 | ||||
NM_134470.3 | Intron | NP_608273.1 | ||||
XM_017006347.1 | Intron | XP_016861836.1 | ||||
XM_017006348.1 | Intron | XP_016861837.1 |