Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 158378 | ||||||||||||||||||||
Literature Links: |
SLC20A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC20A2 - solute carrier family 20 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMIM19 - small integral membrane protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135674.1 | Intron | NP_001129146.1 | ||||
NM_001135675.1 | Intron | NP_001129147.1 | ||||
NM_001135676.1 | Intron | NP_001129148.1 | ||||
NM_138436.3 | Intron | NP_612445.2 | ||||
XM_005273398.4 | Intron | XP_005273455.1 |