Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614694 MIM: 614425 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPRD1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RPRD1B - regulation of nuclear pre-mRNA domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021215.3 | Intron | NP_067038.1 | ||||
XM_005260477.3 | Intron | XP_005260534.1 | ||||
XM_005260480.4 | Intron | XP_005260537.1 | ||||
XM_011528954.2 | Intron | XP_011527256.1 | ||||
XM_017027990.1 | Intron | XP_016883479.1 | ||||
XM_017027991.1 | Intron | XP_016883480.1 |
TTI1 - TELO2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |