Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGAGTATTGTCCTTTGAGGTCAC[G/T]TCAGGTCTTTCTGGTACTAAAACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182396 MIM: 600914 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100289511 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100289511 - uncharacterized LOC100289511 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC10A1 - solute carrier family 10 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003049.3 | Intron | NP_003040.1 |
SRSF5 - serine and arginine rich splicing factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |