Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604224 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARPC2 - actin related protein 2/3 complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005731.3 | Intron | NP_005722.1 | ||||
NM_152862.2 | Intron | NP_690601.1 | ||||
XM_011510471.1 | Intron | XP_011508773.1 | ||||
XM_017003113.1 | Intron | XP_016858602.1 | ||||
XM_017003114.1 | Intron | XP_016858603.1 | ||||
XM_017003115.1 | Intron | XP_016858604.1 |