Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGATTCTAAGGATTGCTTTTGC[A/G]GAGACAGCATGAGCAAAATGGGGCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600039 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2L1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCL2L1 - BCL2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191.3 | Intron | NP_001182.1 | ||||
NM_001317919.1 | Intron | NP_001304848.1 | ||||
NM_001317920.1 | Intron | NP_001304849.1 | ||||
NM_001317921.1 | Intron | NP_001304850.1 | ||||
NM_001322239.1 | Intron | NP_001309168.1 | ||||
NM_001322240.1 | Intron | NP_001309169.1 | ||||
NM_001322242.1 | Intron | NP_001309171.1 | ||||
NM_138578.2 | Intron | NP_612815.1 | ||||
XM_011528964.2 | Intron | XP_011527266.1 | ||||
XM_017027993.1 | Intron | XP_016883482.1 |