Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609492 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RASSF2 - Ras association domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014737.2 | 2692 | UTR 3 | NP_055552.1 | |||
NM_170774.1 | 2692 | UTR 3 | NP_739580.1 | |||
XM_005260895.3 | 2692 | UTR 3 | XP_005260952.1 | |||
XM_011529410.1 | 2692 | UTR 3 | XP_011527712.1 | |||
XM_011529411.1 | 2692 | UTR 3 | XP_011527713.1 | |||
XM_011529412.2 | 2692 | UTR 3 | XP_011527714.1 | |||
XM_017028149.1 | 2692 | UTR 3 | XP_016883638.1 | |||
XM_017028150.1 | 2692 | UTR 3 | XP_016883639.1 | |||
XM_017028151.1 | 2692 | UTR 3 | XP_016883640.1 | |||
XM_017028152.1 | 2692 | UTR 3 | XP_016883641.1 | |||
XM_017028153.1 | 2692 | UTR 3 | XP_016883642.1 |
Set Membership: |
HapMap |