Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGAGCTTCAGCTGTGCGCTTTA[A/G]ATTTTGACGTACAATGTTTTTACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605016 | ||||||||||||||||||||
Literature Links: |
OR2D3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR2D3 - olfactory receptor family 2 subfamily D member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF215 - zinc finger protein 215 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013250.2 | Intron | NP_037382.2 | ||||
XM_006718311.2 | Intron | XP_006718374.1 |