Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605540 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PAPD5 - PAP associated domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040284.2 | Intron | NP_001035374.2 | ||||
NM_001040285.2 | Intron | NP_001035375.2 | ||||
XM_005256092.4 | Intron | XP_005256149.1 | ||||
XM_005256093.4 | Intron | XP_005256150.2 | ||||
XM_011523275.2 | Intron | XP_011521577.1 | ||||
XM_011523276.2 | Intron | XP_011521578.1 | ||||
XM_011523278.2 | Intron | XP_011521580.1 | ||||
XM_017023555.1 | Intron | XP_016879044.1 |
Set Membership: |
HapMap |