Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCCAAAATTATATTTAATAGTG[A/G]GAGACTGGATTCTTTCCCCCTAAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD44 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD44 - ankyrin repeat domain 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195144.1 | Intron | NP_001182073.1 | ||||
NM_153697.2 | Intron | NP_710181.2 | ||||
XM_005246945.1 | Intron | XP_005247002.1 | ||||
XM_005246946.1 | Intron | XP_005247003.1 | ||||
XM_005246947.2 | Intron | XP_005247004.1 | ||||
XM_005246948.2 | Intron | XP_005247005.1 | ||||
XM_006712838.3 | Intron | XP_006712901.1 | ||||
XM_011512148.2 | Intron | XP_011510450.1 | ||||
XM_017005252.1 | Intron | XP_016860741.1 |
Set Membership: |
HapMap |