Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600308 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP4 - aquaporin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317384.1 | 1414 | UTR 3 | NP_001304313.1 | |||
NM_001317387.1 | 1414 | UTR 3 | NP_001304316.1 | |||
NM_001650.5 | 1414 | UTR 3 | NP_001641.1 | |||
NM_004028.4 | 1414 | UTR 3 | NP_004019.1 | |||
XM_011525942.2 | 1414 | UTR 3 | XP_011524244.1 |
AQP4-AS1 - AQP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |