Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601141 MIM: 606213 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNAB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNAB1 - potassium voltage-gated channel subfamily A member regulatory beta subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308217.1 | 3169 | Intron | NP_001295146.1 | |||
NM_001308222.1 | 3169 | Intron | NP_001295151.1 | |||
NM_003471.3 | 3169 | Intron | NP_003462.2 | |||
NM_172159.3 | 3169 | Intron | NP_751891.1 | |||
NM_172160.2 | 3169 | Intron | NP_751892.1 | |||
XM_011513115.2 | 3169 | Intron | XP_011511417.1 | |||
XM_011513116.2 | 3169 | Intron | XP_011511418.1 | |||
XM_011513117.2 | 3169 | Intron | XP_011511419.1 | |||
XM_017007171.1 | 3169 | Intron | XP_016862660.1 | |||
XM_017007172.1 | 3169 | Intron | XP_016862661.1 | |||
XM_017007173.1 | 3169 | Intron | XP_016862662.1 | |||
XM_017007174.1 | 3169 | Intron | XP_016862663.1 |
SSR3 - signal sequence receptor subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308197.1 | 3169 | UTR 3 | NP_001295126.1 | |||
NM_001308204.1 | 3169 | UTR 3 | NP_001295133.1 | |||
NM_001308205.1 | 3169 | UTR 3 | NP_001295134.1 | |||
NM_007107.4 | 3169 | UTR 3 | NP_009038.1 |