Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602886 MIM: 610450 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR39 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPR39 - G protein-coupled receptor 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001508.2 | 2417 | Intron | NP_001499.1 | |||
XM_011511021.2 | 2417 | Intron | XP_011509323.1 |
LYPD1 - LY6/PLAUR domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077427.3 | 2417 | UTR 3 | NP_001070895.1 | |||
NM_001321234.1 | 2417 | UTR 3 | NP_001308163.1 | |||
NM_001321235.1 | 2417 | UTR 3 | NP_001308164.1 | |||
NM_144586.6 | 2417 | UTR 3 | NP_653187.3 | |||
XM_011510546.2 | 2417 | UTR 3 | XP_011508848.1 |
Set Membership: |
HapMap |