Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCAAATGAACATTATTTACATC[G/T]TACATTCTGCTCCTGAAGACAGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189964 MIM: 601558 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2E2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GTF2E2 - general transcription factor IIE subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002095.4 | Intron | NP_002086.1 | ||||
XM_011544509.2 | Intron | XP_011542811.1 | ||||
XM_011544510.1 | Intron | XP_011542812.1 | ||||
XM_017013363.1 | Intron | XP_016868852.1 | ||||
XM_017013364.1 | Intron | XP_016868853.1 |
RBPMS - RNA binding protein with multiple splicing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |