Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCCTTAAAAACCTAATTAAAGT[C/T]TTGGTGATCCCATAAATACGTAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF680 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF680 - zinc finger protein 680 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130022.1 | Intron | NP_001123494.1 | ||||
NM_178558.4 | Intron | NP_848653.2 | ||||
XM_006715959.2 | Intron | XP_006716022.1 | ||||
XM_017012126.1 | Intron | XP_016867615.1 | ||||
XM_017012127.1 | Intron | XP_016867616.1 | ||||
XM_017012128.1 | Intron | XP_016867617.1 | ||||
XM_017012129.1 | Intron | XP_016867618.1 |