Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616177 MIM: 147520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDRGK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDRGK1 - DDRGK domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPA - inosine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267623.1 | Intron | NP_001254552.1 | ||||
NM_001324236.1 | Intron | NP_001311165.1 | ||||
NM_001324237.1 | Intron | NP_001311166.1 | ||||
NM_001324238.1 | Intron | NP_001311167.1 | ||||
NM_001324240.1 | Intron | NP_001311169.1 | ||||
NM_033453.3 | Intron | NP_258412.1 | ||||
NM_181493.3 | Intron | NP_852470.1 | ||||
XM_006723564.3 | Intron | XP_006723627.1 | ||||
XM_006723565.3 | Intron | XP_006723628.1 | ||||
XM_011529234.2 | Intron | XP_011527536.1 |