Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAGAGATTTAGAAATCTTTTCA[A/G]AGTAACCAGCTGAATGTCAGAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CEACAM21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEACAM21 - carcinoembryonic antigen related cell adhesion molecule 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098506.3 | 478 | Intron | NP_001091976.3 | |||
NM_001288773.2 | 478 | Intron | NP_001275702.2 | |||
NM_001290113.1 | 478 | Intron | NP_001277042.1 | |||
NM_033543.5 | 478 | Intron | NP_291021.4 | |||
XM_005278396.4 | 478 | Intron | XP_005278453.1 | |||
XM_005278397.3 | 478 | Intron | XP_005278454.1 | |||
XM_011527450.1 | 478 | Intron | XP_011525752.1 | |||
XM_011527452.1 | 478 | Intron | XP_011525754.1 | |||
XM_017027430.1 | 478 | Intron | XP_016882919.1 | |||
XM_017027431.1 | 478 | Intron | XP_016882920.1 | |||
XM_017027432.1 | 478 | Intron | XP_016882921.1 | |||
XM_017027433.1 | 478 | Intron | XP_016882922.1 | |||
XM_017027434.1 | 478 | Intron | XP_016882923.1 | |||
XM_017027435.1 | 478 | Intron | XP_016882924.1 | |||
XM_017027436.1 | 478 | UTR 5 | XP_016882925.1 | |||
XM_017027437.1 | 478 | Intron | XP_016882926.1 |