Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGGACTGGAGAAGAATTTGGG[C/T]GTGATCAAACTATGGTTGGGAATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609976 MIM: 600658 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIF3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIF3A - hypoxia inducible factor 3 alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP5C - protein phosphatase 5 catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204284.1 | Intron | NP_001191213.1 | ||||
NM_006247.3 | Intron | NP_006238.1 | ||||
XM_017026935.1 | Intron | XP_016882424.1 |
Set Membership: |
HapMap |