Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAAGGGATAATGTGATGTCTTA[C/T]GGGTAAAAAAATTATAAAACACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TC2N PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TC2N - tandem C2 domains, nuclear | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128595.2 | 2244 | UTR 3 | NP_001122067.1 | |||
NM_001128596.2 | 2244 | UTR 3 | NP_001122068.1 | |||
NM_001289134.1 | 2244 | UTR 3 | NP_001276063.1 | |||
NM_152332.5 | 2244 | UTR 3 | NP_689545.1 |