Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGTGCATCTCAATTCAAACTAG[C/T]CATATTTTAGCTGTGCAGTAATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600951 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TERF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TERF1 - telomeric repeat binding factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003218.3 | Intron | NP_003209.2 | ||||
NM_017489.2 | Intron | NP_059523.2 | ||||
XM_005251291.3 | Intron | XP_005251348.1 | ||||
XM_005251292.3 | Intron | XP_005251349.1 | ||||
XM_011517582.2 | Intron | XP_011515884.1 | ||||
XM_017013792.1 | Intron | XP_016869281.1 |