Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTCTTTTTATAGGATAACATAA[C/T]CTCCAACATGTAGTTTACTAAATTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC148 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC148 - coiled-coil domain containing 148 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301684.1 | 2307 | UTR 3 | NP_001288613.1 | |||
NM_001301685.1 | 2307 | Intron | NP_001288614.1 | |||
NM_138803.3 | 2307 | UTR 3 | NP_620158.3 | |||
XM_005246319.3 | 2307 | Intron | XP_005246376.1 | |||
XM_011510638.2 | 2307 | UTR 3 | XP_011508940.1 | |||
XM_017003392.1 | 2307 | UTR 3 | XP_016858881.1 | |||
XM_017003393.1 | 2307 | UTR 3 | XP_016858882.1 | |||
XM_017003394.1 | 2307 | UTR 3 | XP_016858883.1 | |||
XM_017003395.1 | 2307 | UTR 3 | XP_016858884.1 | |||
XM_017003396.1 | 2307 | UTR 3 | XP_016858885.1 | |||
XM_017003397.1 | 2307 | UTR 3 | XP_016858886.1 | |||
XM_017003398.1 | 2307 | UTR 3 | XP_016858887.1 |
CCDC148-AS1 - CCDC148 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |