Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608698 MIM: 607156 | ||||||||||||||||||||
Literature Links: |
DCBLD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCBLD2 - discoidin, CUB and LCCL domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080927.3 | 4162 | UTR 3 | NP_563615.3 | |||
XM_011512419.2 | 4162 | UTR 3 | XP_011510721.1 |
ST3GAL6 - ST3 beta-galactoside alpha-2,3-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |