Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608685 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM118A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM118A - family with sequence similarity 118 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMC1B - structural maintenance of chromosomes 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291501.1 | Intron | NP_001278430.1 | ||||
NM_148674.4 | Intron | NP_683515.4 | ||||
XM_011530144.2 | Intron | XP_011528446.1 | ||||
XM_011530145.2 | Intron | XP_011528447.1 |