Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615854 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMOX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMOX - spermine oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270691.1 | Intron | NP_001257620.1 | ||||
NM_175839.2 | Intron | NP_787033.1 | ||||
NM_175840.2 | Intron | NP_787034.1 | ||||
NM_175841.2 | Intron | NP_787035.1 | ||||
NM_175842.2 | Intron | NP_787036.1 | ||||
XM_011529261.2 | Intron | XP_011527563.1 |
Set Membership: |
HapMap |