Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTGAAATTGTCATGGAAAAAGA[C/T]AGGTCATGGTTAATAAGTAAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600639 MIM: 118425 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CASP2 - caspase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLCN1 - chloride voltage-gated channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000083.2 | Intron | NP_000074.2 | ||||
XM_011515782.2 | Intron | XP_011514084.1 | ||||
XM_017011739.1 | Intron | XP_016867228.1 | ||||
XM_017011740.1 | Intron | XP_016867229.1 |
Set Membership: |
HapMap |