Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD2B - major facilitator superfamily domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080473.1 | Intron | NP_001073942.1 | ||||
XM_005264301.4 | Intron | XP_005264358.1 | ||||
XM_017004063.1 | Intron | XP_016859552.1 | ||||
XM_017004064.1 | Intron | XP_016859553.1 | ||||
XM_017004065.1 | Intron | XP_016859554.1 | ||||
XM_017004066.1 | Intron | XP_016859555.1 | ||||
XM_017004067.1 | Intron | XP_016859556.1 | ||||
XM_017004068.1 | Intron | XP_016859557.1 | ||||
XM_017004069.1 | Intron | XP_016859558.1 | ||||
XM_017004070.1 | Intron | XP_016859559.1 |