Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300390 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHM PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHM - CHM, Rab escort protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000390.3 | 4977 | UTR 3 | NP_000381.1 | |||
NM_001145414.3 | 4977 | Intron | NP_001138886.1 | |||
NM_001320959.1 | 4977 | UTR 3 | NP_001307888.1 | |||
XM_017029242.1 | 4977 | Intron | XP_016884731.1 | |||
XM_017029243.1 | 4977 | UTR 3 | XP_016884732.1 | |||
XM_017029244.1 | 4977 | UTR 3 | XP_016884733.1 | |||
XM_017029245.1 | 4977 | UTR 3 | XP_016884734.1 | |||
XM_017029246.1 | 4977 | UTR 3 | XP_016884735.1 |
Set Membership: |
HapMap |