Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614041 MIM: 603524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RB1 - RB transcriptional corepressor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCBTB2 - RCC1 and BTB domain containing protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001268.3 | 1950 | UTR 3 | NP_001259.1 | |||
NM_001286830.1 | 1950 | UTR 3 | NP_001273759.1 | |||
NM_001286831.1 | 1950 | UTR 3 | NP_001273760.1 | |||
NM_001286832.1 | 1950 | UTR 3 | NP_001273761.1 | |||
XM_005266242.2 | 1950 | UTR 3 | XP_005266299.1 | |||
XM_005266244.2 | 1950 | UTR 3 | XP_005266301.1 | |||
XM_006719759.3 | 1950 | UTR 3 | XP_006719822.1 | |||
XM_011534894.2 | 1950 | UTR 3 | XP_011533196.1 | |||
XM_011534895.1 | 1950 | UTR 3 | XP_011533197.1 | |||
XM_017020369.1 | 1950 | UTR 3 | XP_016875858.1 | |||
XM_017020370.1 | 1950 | UTR 3 | XP_016875859.1 | |||
XM_017020371.1 | 1950 | UTR 3 | XP_016875860.1 | |||
XM_017020372.1 | 1950 | UTR 3 | XP_016875861.1 | |||
XM_017020373.1 | 1950 | UTR 3 | XP_016875862.1 | |||
XM_017020374.1 | 1950 | Intron | XP_016875863.1 | |||
XM_017020375.1 | 1950 | Intron | XP_016875864.1 | |||
XM_017020376.1 | 1950 | Intron | XP_016875865.1 | |||
XM_017020377.1 | 1950 | Intron | XP_016875866.1 | |||
XM_017020378.1 | 1950 | UTR 3 | XP_016875867.1 | |||
XM_017020379.1 | 1950 | UTR 3 | XP_016875868.1 |