Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611797 | ||||||||||||||||||||
Literature Links: |
UQCC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UQCC1 - ubiquinol-cytochrome c reductase complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184977.1 | Intron | NP_001171906.1 | ||||
NM_018244.4 | Intron | NP_060714.3 | ||||
NM_199487.2 | Intron | NP_955781.2 | ||||
XM_011528877.1 | Intron | XP_011527179.1 | ||||
XM_011528878.1 | Intron | XP_011527180.1 | ||||
XM_011528879.1 | Intron | XP_011527181.1 | ||||
XM_011528880.1 | Intron | XP_011527182.1 | ||||
XM_011528881.2 | Intron | XP_011527183.1 | ||||
XM_011528882.1 | Intron | XP_011527184.1 | ||||
XM_011528883.1 | Intron | XP_011527185.1 | ||||
XM_011528884.1 | Intron | XP_011527186.1 |