Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTCTAGAGATCGCCTGCTCAG[A/G]GGGCTCAGATCCCTTGTCATATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164005 | ||||||||||||||||||||
Literature Links: |
NFIX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFIX - nuclear factor I X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271043.2 | Intron | NP_001257972.1 | ||||
NM_001271044.2 | Intron | NP_001257973.1 | ||||
NM_002501.3 | Intron | NP_002492.2 | ||||
XM_005259917.4 | Intron | XP_005259974.1 | ||||
XM_005259918.4 | Intron | XP_005259975.1 | ||||
XM_005259919.3 | Intron | XP_005259976.1 | ||||
XM_005259920.3 | Intron | XP_005259977.1 | ||||
XM_006722760.2 | Intron | XP_006722823.1 | ||||
XM_011528040.1 | Intron | XP_011526342.1 | ||||
XM_017026837.1 | Intron | XP_016882326.1 |