Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACATTTCCACGAGACAAAAGGAA[A/G]AACACAAGAAGACGTCCATAAGGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114212 MIM: 603327 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAPS - calcyphosine | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RANBP3 - RAN binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300865.1 | Intron | NP_001287794.1 | ||||
NM_003624.2 | Intron | NP_003615.2 | ||||
NM_007320.2 | Intron | NP_015559.2 | ||||
NM_007322.2 | Intron | NP_015561.1 | ||||
XM_006722928.1 | Intron | XP_006722991.1 | ||||
XM_006722929.1 | Intron | XP_006722992.1 | ||||
XM_011528391.1 | Intron | XP_011526693.1 | ||||
XM_011528392.1 | Intron | XP_011526694.1 | ||||
XM_011528393.1 | Intron | XP_011526695.1 | ||||
XM_011528394.1 | Intron | XP_011526696.1 | ||||
XM_017027405.1 | Intron | XP_016882894.1 | ||||
XM_017027406.1 | Intron | XP_016882895.1 |
VMAC - vimentin-type intermediate filament associated coiled-coil protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |