Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142620 MIM: 115441 MIM: 610433 MIM: 610434 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD16A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD16A - abhydrolase domain containing 16A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CSNK2B - casein kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G5B - lymphocyte antigen 6 complex, locus G5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G5C - lymphocyte antigen 6 complex, locus G5C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025262.3 | Intron | NP_079538.3 |