Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615708 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF451 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF451 - zinc finger protein 451 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031623.2 | Intron | NP_001026794.1 | ||||
NM_001257273.1 | Intron | NP_001244202.1 | ||||
NM_015555.2 | Intron | NP_056370.2 | ||||
XM_005248994.1 | Intron | XP_005249051.1 | ||||
XM_011514460.1 | Intron | XP_011512762.1 | ||||
XM_011514461.1 | Intron | XP_011512763.1 | ||||
XM_011514462.2 | Intron | XP_011512764.1 | ||||
XM_011514463.2 | Intron | XP_011512765.1 |
Set Membership: |
HapMap |