Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608933 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf49 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C8orf49 - chromosome 8 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEIL2 - nei like DNA glycosylase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135746.1 | 472 | Intron | NP_001129218.1 | |||
NM_001135747.1 | 472 | Intron | NP_001129219.1 | |||
NM_001135748.1 | 472 | Intron | NP_001129220.1 | |||
NM_145043.2 | 472 | UTR 5 | NP_659480.1 | |||
XM_005272382.2 | 472 | Intron | XP_005272439.1 | |||
XM_005272383.2 | 472 | UTR 5 | XP_005272440.1 | |||
XM_017013301.1 | 472 | UTR 5 | XP_016868790.1 |
Set Membership: |
HapMap |