Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAACTGCCTACAGTTGAATGCCT[A/G]CTGTCTGATTGATTGATTGATTGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602846 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2K PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UBE2K - ubiquitin conjugating enzyme E2 K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111112.1 | Intron | NP_001104582.1 | ||||
NM_001111113.1 | Intron | NP_001104583.1 | ||||
NM_001312646.1 | Intron | NP_001299575.1 | ||||
NM_001312647.1 | Intron | NP_001299576.1 | ||||
NM_001312648.1 | Intron | NP_001299577.1 | ||||
NM_005339.4 | Intron | NP_005330.1 |
Set Membership: |
HapMap |