Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATACATCCAGATAGTGGAATACT[C/T]TGAAATTGTAAAAAAAAAAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602136 | ||||||||||||||||||||||||||||||||
Literature Links: |
PEX1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PEX1 - peroxisomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM48 - RNA binding motif protein 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032120.3 | Intron | NP_115496.2 | ||||
XM_005250636.4 | Intron | XP_005250693.1 | ||||
XM_006716149.3 | Intron | XP_006716212.1 | ||||
XM_006716150.3 | Intron | XP_006716213.1 | ||||
XM_017012709.1 | Intron | XP_016868198.1 |
Set Membership: |
HapMap |