Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610025 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL24A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COL24A1 - collagen type XXIV alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152890.5 | 7295 | Intron | NP_690850.2 | |||
XM_017000924.1 | 7295 | Intron | XP_016856413.1 | |||
XM_017000925.1 | 7295 | Intron | XP_016856414.1 | |||
XM_017000926.1 | 7295 | UTR 3 | XP_016856415.1 | |||
XM_017000927.1 | 7295 | UTR 3 | XP_016856416.1 | |||
XM_017000928.1 | 7295 | UTR 3 | XP_016856417.1 | |||
XM_017000929.1 | 7295 | UTR 3 | XP_016856418.1 | |||
XM_017000930.1 | 7295 | Intron | XP_016856419.1 |
Set Membership: |
HapMap |