Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTATTCTGTATACTCTTACTTA[C/T]TGATGTTTCTGCATGAAACTCGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602356 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAF5 - TNF receptor associated factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033910.2 | Intron | NP_001029082.1 | ||||
NM_001319207.1 | Intron | NP_001306136.1 | ||||
NM_004619.3 | Intron | NP_004610.1 | ||||
NM_145759.2 | Intron | NP_665702.1 | ||||
XM_011509957.2 | Intron | XP_011508259.1 | ||||
XM_011509959.2 | Intron | XP_011508261.1 | ||||
XM_011509960.2 | Intron | XP_011508262.1 | ||||
XM_017002221.1 | Intron | XP_016857710.1 |
Set Membership: |
HapMap |