Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAP1 - adenylate cyclase associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105530.1 | Intron | NP_001099000.1 | ||||
NM_006367.3 | Intron | NP_006358.1 | ||||
XM_005270368.1 | Intron | XP_005270425.1 | ||||
XM_011540509.1 | Intron | XP_011538811.1 | ||||
XM_011540510.1 | Intron | XP_011538812.1 | ||||
XM_011540511.1 | Intron | XP_011538813.1 | ||||
XM_011540512.1 | Intron | XP_011538814.1 | ||||
XM_011540513.2 | Intron | XP_011538815.1 | ||||
XM_011540514.1 | Intron | XP_011538816.1 | ||||
XM_011540515.1 | Intron | XP_011538817.1 | ||||
XM_017000067.1 | Intron | XP_016855556.1 | ||||
XM_017000068.1 | Intron | XP_016855557.1 | ||||
XM_017000069.1 | Intron | XP_016855558.1 | ||||
XM_017000070.1 | Intron | XP_016855559.1 | ||||
XM_017000071.1 | Intron | XP_016855560.1 | ||||
XM_017000072.1 | Intron | XP_016855561.1 | ||||
XM_017000073.1 | Intron | XP_016855562.1 |
Set Membership: |
HapMap |