Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTGTGAGACAGCAATGGTGCT[A/G]GGTATCAGGCCCTCTTAGGTCCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610861 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00341 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC00341 - long intergenic non-protein coding RNA 341 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNE3 - spectrin repeat containing nuclear envelope family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152592.3 | 4483 | Intron | NP_689805.3 | |||
XM_005267377.3 | 4483 | UTR 3 | XP_005267434.1 | |||
XM_006720063.3 | 4483 | UTR 3 | XP_006720126.1 | |||
XM_011536515.2 | 4483 | UTR 3 | XP_011534817.1 | |||
XM_017021056.1 | 4483 | UTR 3 | XP_016876545.1 | |||
XM_017021057.1 | 4483 | Intron | XP_016876546.1 |