Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615115 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASXL3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASXL3 - additional sex combs like 3, transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030632.2 | Intron | NP_085135.1 | ||||
XM_005258356.1 | Intron | XP_005258413.1 | ||||
XM_011526205.2 | Intron | XP_011524507.1 | ||||
XM_011526206.2 | Intron | XP_011524508.1 | ||||
XM_011526209.1 | Intron | XP_011524511.1 | ||||
XM_011526210.1 | Intron | XP_011524512.1 | ||||
XM_011526212.1 | Intron | XP_011524514.1 | ||||
XM_011526213.1 | Intron | XP_011524515.1 | ||||
XM_017026012.1 | Intron | XP_016881501.1 | ||||
XM_017026013.1 | Intron | XP_016881502.1 | ||||
XM_017026014.1 | Intron | XP_016881503.1 |